(R)-2-Amino-3-(tritylthio)propanamide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Cys(Trt)-NH2, H-Cys(Trt)-NH₂, S-Trityl-L-cysteinamide

H63628-03 H63628-06
AAH63628-03EA 46.4 USD
AAH63628-03 AAH63628-06
(R)-2-Amino-3-(tritylthio)propanamide 98%
(R)-2-Amino-3-(tritylthio)propanamide

Purity: 98% CAS No.: 166737-85-5 Molecular Formula: C22H22N2OS Formula Weight: 362.49 Physical Form: Powder Appearance: Off-white MDL No.: MFCD22126061

Formula: C₂₂H₂₂N₂OS
MW: 362.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22126061
CAS Number: 166737-85-5

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR