DL-Phenylalaninol 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
DL-Phenylalaninol, 96%
Purity: 96%
CAS No.: 16088-07-6
Molecular Formula: C9H13NO
Formula Weight: 151.21
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 69-73°
Beilstein Reference: 2208237
UN No.: 3259
MDL No.: MFCD00066689
Formula:
C₉H₁₃NO Melting Pt: 69…73 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066689 CAS Number: 16088-07-6 UN: 3259 ADR: "8",III |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...