4-Thiomorpholinoaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63360-03
H63360-MD
AAH63360-03EA
184
USD
AAH63360-03
AAH63360-06
AAH63360-MD
4-Thiomorpholinoaniline 97%
4-Thiomorpholinoaniline
4-(4-Thiomorpholinyl)aniline, 97%
Purity: 97%
CAS No.: 22589-35-1
Molecular Formula: C10H14N2S
Formula Weight: 194.3
Storage Temperature: 2°C to 8°C
Physical Form: Solid
UN No.: 2811
MDL No.: MFCD01625958
Formula:
C₁₀H₁₄N₂S Storage Temperature: Ambient |
MDL Number:
MFCD01625958 CAS Number: 22589-35-1 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...