DL-Glutamine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-DL-Gln-OH, 2,5-Diamino-5-oxopentanoic acid
H62915-03
H62915-06
AAH62915-03EA
53.6
USD
AAH62915-03
AAH62915-06
DL-Glutamine 98%
DL-Glutamine
DL-Glutamine, 98%
Purity: 98%
CAS No.: 585-21-7
Molecular Formula: C5H10N2O
Formula Weight: 146.14
Melting Point: 177°
MDL No.: MFCD00065103
Formula:
C₅H₁₀N₂O₃ Melting Pt: 177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065103 CAS Number: 6899-04-3 Merck Index: 14,4471 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...