cis-4-Isopropylcyclohexanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H62594-03 H62594-06 H62594-MD
AAH62594-03EA 288 USD
AAH62594-03 AAH62594-06 AAH62594-MD
cis-4-Isopropylcyclohexanecarboxylic acid 97%
cis-4-Isopropylcyclohexanecarboxylic acid

Purity: 97% CAS No.: 7084-93-7 Molecular Formula: C10H18O2 Formula Weight: 170.24 Physical Form: Solid

Formula: C₁₀H₁₈O₂
MW: 170.25
Storage Temperature: Ambient
CAS Number: 7084-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR