3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
3-Fluoro-4-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%
Purity: 96%
CAS No.: 1073354-74-1
Molecular Formula: C17H25BFNO3
Formula Weight: 321.19
Physical Form: Solid
Melting Point: 35-40°
MDL No.: MFCD09027076
Formula:
C₁₇H₂₅BFNO₃ Melting Pt: 35…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD09027076 CAS Number: 1073354-74-1 |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...