cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H62281-03
AAH62281-03EA
47.7
USD
AAH62281-03
AAH62281-MD
cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 97%
cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
cis-3-(Boc-amino)cyclohexanecarboxylic acid, 97%
Purity: 97%
CAS No.: 222530-33-8
Molecular Formula: C12H21NO4
Formula Weight: 243.29
Melting Point: 143-147°
MDL No.: MFCD01862295
Formula:
C₁₂H₂₁NO₄ Melting Pt: 143…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD01862295 CAS Number: 222530-33-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...