1-(Boc-amino)cyclopropanecarboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(Boc-Amino)cyclopropanecarboxylic acid, 1-Aminocyclopropane-1-carboxylic acid, N-BOC protected, 1-[(tert-Butoxycarbonyl)amino]cyclopropanecarboxylic acid
H61992-03
AAH61992-03EA
86.8
USD
AAH61992-03
AAH61992-MD
1-(Boc-amino)cyclopropanecarboxylic acid 98%
1-(Boc-amino)cyclopropanecarboxylic acid
1-(Boc-amino)cyclopropanecarboxylic acid, 98%
Purity: 98%
CAS No.: 88950-64-5
Molecular Formula: C9H15NO4
Formula Weight: 201.22
Melting Point: 118° dec
Beilstein Reference: 4430628
MDL No.: MFCD00083257
Formula:
C9H15NO4 MW: 201.22 g/mol Melting Pt: 118 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00083257 CAS Number: 88950-64-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...