(±)-2-Hydroxyisovaleric acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H60872-03
AAH60872-03EA 77.6 USD
AAH60872-03
(±)-2-Hydroxyisovaleric acid 98%
(±)-2-Hydroxyisovaleric acid

CAS: 4026-18-0 MDL: MFCD00004242

Formula: C₅H₁₀O₃
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00004242
CAS Number: 4026-18-0
EINECS: 223-697-5

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR