(5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 98%
CAS No.: 100516-54-9
Molecular Formula: C24H21NO4
Formula Weight: 387.43
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 205-207°
Optical Rotation: -66° (c=5.5 in methylene chloride)
MDL No.: MFCD00074958
Formula:
C₂₄H₂₁NO₄ Melting Pt: 205…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074958 CAS Number: 100516-54-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...