Phenyl bromoacetate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bromoacetic acid Phenyl ester, Phenyl 2-bromoacetate

H60464-06 H60464-18
AAH60464-06EA 34.5 USD
AAH60464-06 AAH60464-18
Phenyl bromoacetate 98%
Phenyl bromoacetate

CAS: 620-72-4 MDL: MFCD00192391

Formula: C₈H₇BrO₂
Boiling Pt: 134 °C
Melting Pt: 31…33 °C
Density: 1.508
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00192391
CAS Number: 620-72-4

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR