(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-DTBM-Garphos, (R)-2,2'-Bis[bis(4-methoxy-3,5-di-t-butylphenyl)phosphino]-4,4',6,6'-tetramethoxy)-1,1'-biphenyl, (R)-(4,4',6,6'-Tetramethoxybiphenyl-2,2'-diyl)bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphine]
H60112-03
H60112-MD
AAH60112-03EA
746
USD
AAH60112-03
AAH60112-MD
(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl ≥97%
(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl
Purity: 97+%
CAS No.: 1365531-98-1
Molecular Formula: C76H108O8P2
Formula Weight: 1211.61
MDL No.: MFCD20265636
Formula:
C₇₆H₁₀₈O₈P₂ MW: 1211.61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20265636 CAS Number: 1365531-98-1 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...