N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Tyr-OH, N-Fmoc-L-tyrosine
H59967-03
AAH59967-03EA
61.1
USD
AAH59967-03
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine 97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine
Purity: 97%
CAS No.: 92954-90-0
Molecular Formula: C24H21NO5
Formula Weight: 403.43
Storage Temperature: 2°C to 8°C
Melting Point: 182-187°
Beilstein Reference: 3573123
Optical Rotation: -20° (c=1 in DMF)
MDL No.: MFCD00134890
Formula:
C₂₄H₂₁NO₅ MW: 403.43 g/mol Melting Pt: 182…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134890 CAS Number: 92954-90-0 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...