1-n-Butyl-3-methylimidazolium di-n-butyl phosphate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59893-06
H59893-18
AAH59893-06EA
53.3
USD
AAH59893-06
AAH59893-18
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate 96%
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate
Purity: 96%
CAS No.: 663199-28-8
Molecular Formula: C16H33N2O4P
Formula Weight: 348.42
Physical Form: Liquid
Refractive Index: 1.4720
Sensitivity: Moisture Sensitive
Appearance: Brown
MDL No.: MFCD09265043
Formula:
C₁₆H₃₃N₂O₄P Storage Temperature: Ambient |
MDL Number:
MFCD09265043 CAS Number: 663199-28-8 |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...