L-(+)-Methyl-tert-leucinate hydrochloride 97%

Supplier: Thermo Scientific Chemicals

H59563-03
AAH59563-03EA 189 USD
AAH59563-03
L-(+)-Methyl-tert-leucinate hydrochloride 97%
L(+)-Methyl tert-leucinate hydrochloride

CAS: 63038-27-7 MDL: MFCD00040604

Formula: C₇H₁₅NO₂·HCl
Melting Pt: 183 °C
Storage Temperature: Ambient
MDL Number: MFCD00040604
CAS Number: 63038-27-7

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR