Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine 97% (by HPLC)
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Asn(Trt)-OH, Nɑ-Fmoc-Nγ-trityl-L-asparagine
H59536-06
H59536-14
AAH59536-06EA
67.9
USD
AAH59536-06
AAH59536-14
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine 97% (by HPLC)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-gamma-trityl-L-asparagine
Purity: 97%
CAS No.: 132388-59-1
Molecular Formula: C38H32N2O5
Formula Weight: 596.67
Storage Temperature: 2°C to 8°C
Melting Point: 201-204°
Beilstein Reference: 4343823
Optical Rotation: -15° (c=1 in methanol)
MDL No.: MFCD00077049
Formula:
C₃₈H₃₂N₂O₅ Melting Pt: 201…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077049 CAS Number: 132388-59-1 |
Specification Test Results
Assay (HPLC) | Min. 96.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...