1-Ethyl-2,3-dimethylimidazolium chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

H59225-06 H59225-18
AAH59225-06EA 82.4 USD
AAH59225-06 AAH59225-18
1-Ethyl-2,3-dimethylimidazolium chloride 97%
1-Ethyl-2,3-dimethylimidazolium chloride

CAS: 92507-97-6 MDL: MFCD03093290

Formula: C₇H₁₃ClN₂
Storage Temperature: Ambient
MDL Number: MFCD03093290
CAS Number: 92507-97-6

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR