4-Iodo-1H-pyrazole 99%

Supplier: Thermo Scientific Chemicals
Warning

H56369-03 H56369-06 H56369-14
AAH56369-03EA 25.7 USD
AAH56369-03 AAH56369-06 AAH56369-14
4-Iodo-1H-pyrazole 99%
4-Iodo-1H-pyrazole

CAS: 3469-69-0 MDL: MFCD00005244

Formula: C₃H₃IN₂
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005244
CAS Number: 3469-69-0
EINECS: 222-434-1

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR