5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) 98+%
Supplier: Thermo Scientific Chemicals
H56265-03
AAH56265-03EA
323
USD
AAH56265-03
AAH56265-06
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) 98+%
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Purity: 98+%
CAS No.: 102185-33-1
Molecular Formula: C8H4BrClNNa2O4P
Formula Weight: 370.43
Storage Temperature: -30°C to -10°C
Formula:
C₈H₄BrClNNa₂O₄P Storage Temperature: Refrigerator |
MDL Number:
MFCD00036757 CAS Number: 102185-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...