N,N'-Bis(2-aminoethyl)-1,3-propanediamine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,7-Diaza-1,9-nonanediamine, N1,N1'-(Propane-1,3-diyl)bis(ethane-1,2-diamine), 1,4,8,11-Tetraazaundecane
H54950-03
AAH54950-03EA
52
USD
AAH54950-03
N,N'-Bis(2-aminoethyl)-1,3-propanediamine 96%
N,N'-Bis(2-aminoethyl)-1,3-propanediamine
Purity: 97%
CAS No.: 4741-99-5
Molecular Formula: C7H20N4
Formula Weight: 160.26
Flash Point: 100°(212°F)
Refractive Index: 1.494
Boiling Point: 142-145°
Beilstein Reference: 1742732
UN No.: 2735
MDL No.: MFCD00008174
Formula:
C₇H₂₀N₄ Boiling Pt: 142…145 °C Density: 0.96 Flash Pt: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008174 CAS Number: 4741-99-5 EINECS: 225-254-1 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...