Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥97%

Supplier: Thermo Scientific Chemicals
Warning

H54807-MB H54807-MC
AAH54807-MBEA 166 USD
AAH54807-MB AAH54807-MC
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥97%
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)

Purity: 98+% CAS No.: 92921-24-9 Molecular Formula: C16H17N2NaO9S Formula Weight: 436.37 Storage Temperature: 2°C to 8°C MDL No.: MFCD00054978

Formula: C₁₆H₁₈N₂O₉S
Storage Temperature: Ambient
MDL Number: MFCD00054978
CAS Number: 92921-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR