2-Ethylhexyl-4-(dimethylamino)benzoate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Ethylhexyl 4-(dimethylamino)benzoate
H54136-AE
H54136-AP
AAH54136-AEEA
53.3
USD
AAH54136-AE
AAH54136-AP
2-Ethylhexyl-4-(dimethylamino)benzoate 99%
2-Ethylhexyl 4-(dimethylamino)benzoate
Purity: 99%
CAS No.: 21245-02-3
Molecular Formula: C17H27NO2
Formula Weight: 277.40
Flash Point: 113°(235°F)
Refractive Index: 1.5420
Boiling Point: 325°
Merck Reference: 14,3232
MDL No.: MFCD00017526
Formula:
C₁₇H₂₇NO₂ MW: 277.40 g/mol Boiling Pt: 325 °C Density: 0.995 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017526 CAS Number: 21245-02-3 EINECS: 244-289-3 Merck Index: 14,3232 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...