N,N'-Bis-Z-1-Guanylpyrazole ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N'-Bis-Z-1-Guanylpyrazole

H54092-03 H54092-06
AAH54092-03EA 83.8 USD
AAH54092-03 AAH54092-06
N,N'-Bis-Z-1-Guanylpyrazole ≥98%
N,N'-Bis-Z-1-guanylpyrazole

Purity: 98+% CAS No.: 152120-55-3 Molecular Formula: C20H18N4O4 Formula Weight: 378.38 Melting Point: 87-91° MDL No.: MFCD02683516

Formula: C₂₀H₁₈N₄O₄
MW: 378.38 g/mol
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD02683516
CAS Number: 152120-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR