1-Phenyl-1-hexyn-3-ol 97%
Supplier: Thermo Scientific Chemicals
H53437-06
H53437-14
AAH53437-06EA
66.4
USD
AAH53437-06
AAH53437-14
1-Phenyl-1-hexyn-3-ol 97%
1-Phenyl-1-hexyn-3-ol
Purity: 97%
CAS No.: 1817-51-2
Molecular Formula: C12H14O
Formula Weight: 174.24
Flash Point: 110°(230°F)
Refractive Index: 1.549
MDL No.: MFCD00046687
Formula:
C₁₂H₁₄O Density: 0.949 Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00046687 CAS Number: 1817-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...