2',3'-O-Isopropylideneadenosine 98%
Supplier: Thermo Scientific Chemicals
H53401-06
H53401-14
AAH53401-06EA
61.4
USD
AAH53401-06
AAH53401-14
2',3'-O-Isopropylideneadenosine 98%
2',3'-O-Isopropylideneadenosine
Purity: 98%
CAS No.: 362-75-4
Molecular Formula: C13H17N5O4
Formula Weight: 307.31
Melting Point: 221-222°
Beilstein Reference: 43435
Optical Rotation: -99° (c=1 in dioxane)
MDL No.: MFCD00005756
Formula:
C₁₃H₁₇N₅O₄ Melting Pt: 221…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005756 CAS Number: 362-75-4 EINECS: 206-650-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...