(2-Methylsulfinylphenyl)boronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53209-MD
AAH53209-03EA
389.99
USD
AAH53209-03
AAH53209-MD
(2-Methylsulfinylphenyl)boronic acid 97%
(2-Methylsulfinylphenyl)boronic acid
Purity: 97%
CAS No.: 850567-97-4
Molecular Formula: C7H9BO3S
Formula Weight: 184.02
Melting Point: 118-122°
MDL No.: MFCD03095261
Formula:
C₇H₉BO₃S Melting Pt: 118…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095261 CAS Number: 850567-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...