4-Fluoro-3-(methylcarbamoyl)phenylboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Fluoro-3-(methylcarbamoyl)phenyl)boronic acid, 4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid

H53101-03 H53101-MD
AAH53101-03EA 402 USD
AAH53101-03 AAH53101-MD
4-Fluoro-3-(methylcarbamoyl)phenylboronic acid 96%
4-Fluoro-3-(methylcarbamoyl)phenylboronic acid

Purity: 96% CAS No.: 874219-19-9 Molecular Formula: C8H9BFNO3 Formula Weight: 196.97 Melting Point: 226-228° MDL No.: MFCD08235038

Formula: C₈H₉BFNO₃
MW: 196.97 g/mol
Melting Pt: 226…228 °C
Storage Temperature: Ambient
MDL Number: MFCD08235038
CAS Number: 874219-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR