(S)-1-(Boc-amino)-2-propanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl (S)-(2-hydroxypropyl)carbamate

H52826-06 H52826-14
AAH52826-06EA 63.2 USD
AAH52826-06 AAH52826-14
(S)-1-(Boc-amino)-2-propanol 97%
(S)-1-(Boc-amino)-2-propanol

Purity: 97% CAS No.: 167938-56-9 Molecular Formula: C8H17NO3 Formula Weight: 175.23 MDL No.: MFCD04974340

Formula: C₈H₁₇NO₃
MW: 175.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974340
CAS Number: 167938-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR