Fmoc-3-(1-naphthyl)-L-alanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-1-Nal-OH
H51967-03
H51967-06
AAH51967-03EA
180
USD
AAH51967-03
AAH51967-06
Fmoc-3-(1-naphthyl)-L-alanine 98%
Fmoc-3-(1-naphthyl)-L-alanine
Purity: 98%
CAS No.: 96402-49-2
Molecular Formula: C28H23NO4
Formula Weight: 437.49
Melting Point: 171-183°
Beilstein Reference: 4829118
MDL No.: MFCD00151914
Formula:
C₂₈H₂₃NO₄ MW: 437.495 g/mol Melting Pt: 171…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151914 CAS Number: 96402-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...