4-(Tributylstannyl)thiazole 95%

Supplier: Thermo Scientific Chemicals
Danger

H51457-03 H51457-MD
AAH51457-03EA 283 USD
AAH51457-03 AAH51457-MD
4-(Tributylstannyl)thiazole 95%
4-(Tributylstannyl)thiazole

Purity: 95% CAS No.: 173979-01-6 Molecular Formula: C15H29NSSn Formula Weight: 374.17 UN No.: 2788 MDL No.: MFCD01319073

Formula: C₁₅H₂₉NSSn
MW: 374.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319073
CAS Number: 173979-01-6
UN: 2788
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR