Cyclohexylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)

Supplier: Thermo Scientific Chemicals
Danger

H51165-AE H51165-AP
AAH51165-AEEA 59.36 USD
AAH51165-AE AAH51165-AP
Cyclohexylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)
Cyclohexylmagnesium chloride

CAS No.: 931-51-1 Molecular Formula: C6H11ClMg Formula Weight: 142.91 UN No.: 2924 MDL No.: MFCD00003816

Formula: C₆H₁₁ClMg
MW: 142.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003816
CAS Number: 931-51-1
UN: 2924
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR