Platinum 5% on carbon paste wetted with ca. 50% water, sulfided B106032-5

Supplier: Thermo Scientific Chemicals
Warning

H36544-06 H36544-14
AAH36544-06EA 128 USD
AAH36544-06 AAH36544-14 AAH36544-22
Platinum 5% on carbon paste wetted with ca. 50% water, sulfided B106032-5
Platinum

Platinum, 5% on carbon, sulfided, B106032-5


CAS No.: 7440-06-4 Physical Form: Solid Appearance: Dark grey to black Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR