3-Iodo-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Iodopyrazole

H36465-03 H36465-06
AAH36465-03EA 58.4 USD
AAH36465-03 AAH36465-06
3-Iodo-1H-pyrazole 97%
3-Iodo-1H-pyrazole

3-Iodo-1H-pyrazole, 97%


CAS: 4522-35-4

Formula: C₃H₃IN₂
MW: 193.98 g/mol
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD12022324
CAS Number: 4522-35-4

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR