Lithium hydroxide monohydrate battery grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
H36379-0B
H36379-30
AAH36379-0BEA
439.44
USD
AAH36379-0B
AAH36379-30
Lithium hydroxide monohydrate battery grade
Lithium hydroxide monohydrate
Lithium hydroxide monohydrate, battery grade
CAS: 1310-66-3
MDL: MFCD00149772
Formula:
LiOH·1H₂O Boiling Pt: 920 °C (1013 hPa) Melting Pt: 462 °C Density: 1.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149772 CAS Number: 1310-66-3 EINECS: 215-183-4 UN: 2680 ADR: 8,II Merck Index: 13,05556 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...