Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
H35916-03
H35916-06
AAH35916-03EA
60.8
USD
AAH35916-03
AAH35916-06
Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
CAS: 353258-31-8
MDL: MFCD01833013
Formula:
C₁₁H₈ClF₃N₂O₂ MW: 292.65 g/mol Melting Pt: 132…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD01833013 CAS Number: 353258-31-8 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...