2-Chloro-1H-indole-3-carbaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H34381-03 H34381-09
AAH34381-03EA 63.3 USD
AAH34381-03 AAH34381-09
2-Chloro-1H-indole-3-carbaldehyde 97%
2-Chloro-1H-indole-3-carbaldehyde

Purity: 97% CAS No.: 5059-30-3 Molecular Formula: C9H6ClNO Formula Weight: 179.61 Melting Point: ca 212° dec. MDL No.: MFCD00111620

Formula: C₉H₆ClNO
MW: 179.61 g/mol
Melting Pt: ∼ 212 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00111620
CAS Number: 5059-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR