Docosanal 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33924-03
H33924-MD
AAH33924-03EA
312.91
USD
AAH33924-03
AAH33924-MD
Docosanal 98%
Docosanal
Purity: 98%
CAS No.: 57402-36-5
Molecular Formula: C22H44O
Formula Weight: 324.59
Melting Point: 57-60°
MDL No.: MFCD07780595
Formula:
C₂₂H₄₄O Melting Pt: 57…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD07780595 CAS Number: 57402-36-5 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...