Hydrogen chloride 3 mol/l in cyclopentyl methyl ether

Supplier: Thermo Scientific Chemicals
Danger

H33398-AE H33398-AP
AAH33398-AEEA 89.02 USD
AAH33398-AE AAH33398-AP
Hydrogen chloride 3 mol/l in cyclopentyl methyl ether
Hydrogen chloride

CAS: 7647-01-0 MDL: MFCD00011324

Formula: HCl
MW: 36.46
Boiling Pt: 106 °C
Density: 0.895
Flash Pt: 10 °C (50 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR