Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]-thiophene-3-carboxylate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-5,6-dihydro-4-[4H]-cyclopenta[B]thiophene-3-carboxylic acid ethyl ester, Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate

H33196-06 H33196-14
AAH33196-06EA 98.8 USD
AAH33196-06 AAH33196-14
Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]-thiophene-3-carboxylate 96%
Ethyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate

Purity: 96% CAS No.: 4815-29-6 Molecular Formula: C10H13NO2S Formula Weight: 211.28 Melting Point: 95-96° MDL No.: MFCD00102063

Formula: C₁₀H₁₃NO₂S
MW: 211.28 g/mol
Melting Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00102063
CAS Number: 4815-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR