Zinc chloride 0.7 M in THF

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

H32962-AE H32962-AP
AAH32962-AEEA 135.96 USD
AAH32962-AE AAH32962-AP
Zinc chloride 0.7 M in THF
Zinc chloride

CAS No.: 7646-85-7 Molecular Formula: Cl2Zn Formula Weight: 136.29 UN No.: 1993 MDL No.: MFCD00011295

Formula: ZnCl₂
Density: 0.948
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2924
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR