Agarose 4% B - CL, cross-linked, beads
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32305-AU
AAH32305-AUEA
1384
USD
AAH32305-AU
Agarose 4% B - CL, cross-linked, beads
Agarose
CAS No.: 61970-08-9
Storage Temperature: 2°C to 8°C
Flash Point: 38°(100°F)
Boiling Point: 100°
UN No.: 1170
MDL No.: MFCD00081294
Boiling Pt:
100 °C Density: 1.040 Flash Pt: 38 °C(100 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00081294 CAS Number: 9012-36-6 UN: 1170 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...