Stains-all 96%
Supplier: Thermo Scientific Chemicals
General protein and nucleic acid stain, which stains different colors on bands of RNA (bluish purple), DNA (blue) and proteins (red)
Purity: 96%
CAS No.: 7423-31-6
Molecular Formula: C30H27BrN2S2
Formula Weight: 559.58
Beilstein Reference: 3865213
MDL No.: MFCD00078788
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...