Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, (H₂O 1-5%) 97% may cont. 1-5% water
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Kobayashi's Reagent, NaBArF24
H30014-14
H30014-03
H30014-06
AAH30014-14EA
1784
USD
AAH30014-14
AAH30014-03
AAH30014-06
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, (H₂O 1-5%) 97% may cont. 1-5% water
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate
Purity: 97%
CAS No.: 79060-88-1
Molecular Formula: C32H12BF24Na
Formula Weight: 886.20
Melting Point: 310° dec.
MDL No.: MFCD00043323
Formula:
C₃₂H₁₂BF₂₄Na Melting Pt: 310 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00043323 CAS Number: 79060-88-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...