(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic Acid, H-D-Tic-OH, D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

H29112-03 H29112-MD H29112-06
AAH29112-03EA 233 USD
AAH29112-03 AAH29112-MD AAH29112-06
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

CAS No.: 103733-65-9 Molecular Formula: C10H11NO2 Formula Weight: 177.20 Optical Rotation: +163° (c=2 in 1N NaOH) MDL No.: MFCD00144038

Formula: C₁₀H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00144038
CAS Number: 103733-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR