Dichloro(dimethylamino)phosphine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethylphosphoramidous dichloride, (Dimethylamino)phosphonous Dichloride
H27856-06
H27856-14
AAH27856-06EA
68.68
USD
AAH27856-06
AAH27856-14
Dichloro(dimethylamino)phosphine 98%
Dichloro(dimethylamino)phosphine
Purity: 98%
CAS No.: 683-85-2
Molecular Formula: C2H6Cl2NP
Formula Weight: 145.96
Storage Temperature: 2°C to 8°C
Flash Point: -23°(-9°F)
Boiling Point: 150°
UN No.: 2924
MDL No.: MFCD00013613
Formula:
C₂H₆Cl₂NP MW: 145.95 g/mol Boiling Pt: 150 °C Density: 1.271 Flash Pt: –23 °C (–9 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013613 CAS Number: 683-85-2 UN: 2924 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...