n-Pentane 99+% for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Pentane
H27427-0F
H27427-AU
AAH27427-0FEA
199.87
USD
AAH27427-0F
AAH27427-AU
n-Pentane 99+% for HPLC
n-Pentane
Purity: 99+%
CAS No.: 109-66-0
Molecular Formula: CH3(CH2)3CH3
Formula Weight: 72.15
Flash Point: -49°(-57°F)
Refractive Index: 1.3580
Odor: Nearly odorless
Melting Point: -130°
Boiling Point: 36°
Merck Reference: 14,7116
UN No.: 1265
MDL No.: MFCD00009498
Formula:
H₃C(CH₂)₃CH₃ MW: 72.15 g/mol Boiling Pt: 36 °C Melting Pt: –130 °C Density: 0.626 Flash Pt: -49 °C(-57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009498 CAS Number: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: "3",II REACH: 01-2119459286-30 Merck Index: 14,7116 |
Specification Test Results
Appearance (Color) | Clear, colorless |
Form | Liquid |
Assay (unspecified) (all isomers) | >99.5% |
Assay (unspecified) (n-Pentane) | >95% |
Water Content (Karl Fischer Titration) | < 50 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...