2-Chloro-D-phenylalanine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-D-Phe(2-Cl)-OH

H27359-03 H27359-MD
AAH27359-03EA 147 USD
AAH27359-03 AAH27359-MD
2-Chloro-D-phenylalanine 98%
2-Chloro-D-phenylalanine

Purity: 98% CAS No.: 80126-50-7 Molecular Formula: C9H10ClNO2 Formula Weight: 199.64 Melting Point: 211-213° MDL No.: MFCD00077920

Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
Melting Pt: 211…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00077920
CAS Number: 80126-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR