5-Bromo-2'-deoxyuridine (broxuridine) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 99%
CAS No.: 59-14-3
Molecular Formula: C9H11BrN2O5
Formula Weight: 307.10
Storage Temperature: 2°C to 8°C
Melting Point: ca 190° dec.
Beilstein Reference: 30395
Merck Reference: 14,1452
Optical Rotation: +31° (c=1 in 0.1N HCl)
MDL No.: MFCD00006529
Formula:
C₉H₁₁BrN₂O₅ Melting Pt: ∼ 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006529 CAS Number: 59-14-3 EINECS: 200-415-9 Merck Index: 14,1452 |
Specification Test Results
Optical rotation | +31° (c=1 in 0.1N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...