2-Bromoisonicotinonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H27191-03 H27191-06
AAH27191-03EA 96.9 USD
AAH27191-03 AAH27191-06
2-Bromoisonicotinonitrile 97%
2-Bromoisonicotinonitrile

Purity: 97% CAS No.: 10386-27-3 Molecular Formula: C6H3BrN2 Formula Weight: 183.01 Odor: Odorless Melting Point: 70-74° MDL No.: MFCD07367879

Formula: C₆H₃BrN₂
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD07367879
CAS Number: 10386-27-3
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR