Boc-trans-4-hydroxy-L-proline 97%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, Boc-Hyp-OH, trans-N-Boc-4-hydroxy-L-proline

H27110-06 H27110-14
AAH27110-03EA 34.25 USD
AAH27110-03 AAH27110-06 AAH27110-14
Boc-trans-4-hydroxy-L-proline 97%
Boc-trans-4-hydroxy-L-proline

Purity: 97% CAS No.: 13726-69-7 Molecular Formula: C10H17NO5 Formula Weight: 231.25 Melting Point: 123-127° MDL No.: MFCD00053370

Formula: C₁₀H₁₇NO₅
MW: 231.3 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00053370
CAS Number: 13726-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR